U-Tube Viscometer
U-Tube Viscometer
Actual product may vary
Fisherbrand
U-Tube Viscometer
Suitable for transparent liquids. With UKAS certificate of calibration
Have Questions?
Catalog NumberViscosity RangeSize
FKS131610230.9 to 3 cStA
Catalog number FKS13161023
Price (MYR)
2,676.00
EA
Add to cart
Viscosity Range:
0.9 to 3 cSt
Size:
A
Price (MYR)
2,676.00
EA
Add to cart
  • PSL calibrated viscometers are supplied with UKAS certificates providing an official assurance of the competence of the calibration work carried out. UKAS certificates of calibration, which satisfy the requirements of ISO 9000, are issued by the PSL Calibration Laboratory, which is a National Measurement Accreditation Service accredited laboratory.
  • Type BS/U, IP71.Suitable for transparent liquids. Nominal overall length 300mm. Approx. sample size 13mL to 40mL.
Specifications
Product TypeU-Tube Viscometer
Viscosity Range0.9 to 3 cSt
Certifications/ComplianceUKAS, ISO 9000
SizeA
Volume (Metric)18 mL
Unit SizeEA