Search Thermo Fisher Scientific
| Catalog Number | Size | Size (Metric) Chest |
|---|---|---|
| FKS13151731 | Large | 106.68 to 111.76 cm |
| FKS13161731 | Medium | 101.6 to 104.14 cm |
| FKS13171731 | Small | 91.44 to 96.52 cm |
| FKS13181731 | X-Large | 116.84 to 121.92 cm |