Check Order Status
Azlon™ Polypropylene Pipette Stand
Actual product may vary
Fisherbrand
Azlon™ Polypropylene Pipette Stand
Constructed of blue, chemically resistant polypropylene. Azlon™ Polypropylene Pipette Stand holds up to 94 pipettes vertically. Rotatable rack is suspended by a support rod.
Have Questions?
Catalog Number
Material
FKS13108596
Polypropylene
Catalog number
FKS13108596
Price (MYR)
1,254.00
PK
Add to cart
Material:
Polypropylene
Price (MYR)
1,254.00
PK
Add to cart
Blue chemically resistant polypropylene stand
Holds up to 94 pipettes vertically
Very stable with 230mm diameter base
Height of 470mm
Has a rotatable rack suspended by a support rod
Specifications
Material
Polypropylene
Height (Metric)
470 mm
Diameter (Metric) Outer
230 mm
Diameter (Metric) Rod
12 mm
Holds
94 Pipettes
Product Type
Pipette stand
Color
Blue
Quantity
1 Pk.