Thermometer Stand
Thermometer Stand
Actual product may vary
Fisherbrand
Thermometer Stand
Supports 18 thermometers in an upright and secure position.
Have Questions?
Catalog NumberMaterial
FKS13078623Aluminum, Stainless Steel
Catalog number FKS13078623
Price (MYR)
383.00
EA
Add to cart
Material:
Aluminum, Stainless Steel
Price (MYR)
383.00
EA
Add to cart
  • With protective foam cushion on the base
  • Manufactured from aluminum
Specifications
Cell Dividers18
Product TypeThermometer Rack
MaterialAluminum, Stainless Steel
Positions18
For Use With (Equipment)Thermometer
Quantity1 Each
Length (Metric)120 mm
Width (Metric)165 mm
Height (Metric)125 mm
Height (Metric) Interior125 mm
Depth (Metric)120 mm
Diameter (Metric) Outer10 mm
Unit SizeEA