Horizontal Fridge Freezer Thermometer
Horizontal Fridge Freezer Thermometer
Actual product may vary
Horizontal Fridge Freezer Thermometer
Horizontal Fridge Freezer Thermometer
Fisherbrand
Horizontal Fridge Freezer Thermometer
Indicates recommended food storage temperature zones. Brannan™ Horizontal Fridge Freezer Thermometer sits on a shelf or hangs from a rack.
Have Questions?
Catalog NumberTemperature Range (Metric)
FKS13049202-30°C to +40°C
Catalog number FKS13049202
Price (MYR)
46.00
EA
Add to cart
Temperature Range (Metric):
-30°C to +40°C
Price (MYR)
46.00
EA
Add to cart
Simple to use white plastic fridge thermometer with a clear blue temperature scale indicates recommended zones.
  • Fridge temperatures are displayed in °C only.
  • Sits on a shelf or hangs from a rack.
  • 130mm Length
  • White
Specifications
Dimensions (L x W x H)130 x 22 x 40 mm
Display Dimensions218 x 95 x 0.5 mm
Height (Metric)40 mm
Length (Metric)130 mm
Thermometer FillRed Spirit
Max. Temperature (Metric)40°C
Min. Temperature (Metric)-30°C
Temperature Range (Metric)-30°C to +40°C
Accuracy±1°C
DisplayBlue Scale
Width (Metric)22 mm
Unit SizeEA