Check Order Status
Dreyers Agglutination Tubes
Actual product may vary
Dreyers Agglutination Tubes
Have Questions?
Catalog Number
Diameter (Metric)
FKS13048423
7 mm
Catalog number
FKS13048423
Price (MYR)
396.00
PK
Add to cart
Diameter (Metric):
7 mm
Price (MYR)
396.00
PK
Add to cart
Specifications
Length (Metric)
60 mm
Material
Borosilicate Glass
Packaging
100
Quantity
100 Pack
Diameter (Metric)
7 mm
Product Type
Dreyers Agglutination Tube