Bio-bins™ Yellow Paper Based Non-Sharps Infectious Clinical Waste Containers for Incineration Only
Bio-bins™ Yellow Paper Based Non-Sharps Infectious Clinical Waste Containers for Incineration Only
Actual product may vary
Fisherbrand
Bio-bins™ Yellow Paper Based Non-Sharps Infectious Clinical Waste Containers for Incineration Only
Use Econix™ Bio-bins™ The Bio-bin™ is a Paper based Non-sharps Container used to dispose of single use implements, tips, petri dishes, pads and other lab waste (excluding needles and blades) and reduce over filling issues for your non-sharp clinical waste. The Bio-bin has a built-in temporary seal, absorbent mat and is waterproof.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1298367530 L
Catalog number FKS12983675
Price (MYR)
1,099.00
PK
Add to cart
Capacity (Metric):
30 L
Price (MYR)
1,099.00
PK
Add to cart
  • 96% paper based non-plastic single-use non-sharps waste container
  • Comes fold-flat for easy storage
  • Rigid, leakproof construction
  • Leakproof, built-in lid
  • Built-in absorbent Mat
  • UN 3291 approved
Includes temporary seal Autoclavable
Specifications
Capacity (Metric)30 L
Certifications/ComplianceUN3291
Closure TypeTemporary and Permanent
DesignSpace efficient fold-flat
ColorYellow
Height (Metric)500 mm closed, 650 mm open
Length (Metric)290 mm
Quantity10 Pk.
SterilityNon-sterile
Width (Metric)290 mm
AutoclavableAutoclavable
For Use With (Application)Infectious Waste
MaterialCardboard
Product TypeNon-Sharps Waste Container
Unit SizePK