Glass Staining Block
Glass Staining Block
Actual product may vary
Fisherbrand
Glass Staining Block
Position up to 20 microscope slides back-to-back in the Glass Staining Block for effective slide staining.The piece of laboratory glassware is made of clear, polished glass with a heavy cover to minimize evaporation.
Have Questions?
Catalog NumberSlide Count
FKS1289873520
Catalog number FKS12898735
Price (MYR)
180.00
EA
Add to cart
Slide Count:
20
Price (MYR)
180.00
EA
Add to cart
  • Clear polished glass
  • Cavity 30mm diameter x 6mm deep
  • Complete with flat glass cover
  • 40mm square to minimize evaporation
Specifications
Closure IncludedClosure Included
Closure MaterialGlass
Bottom ShapeFlat
MaterialGlass
TypeStaining Block
For Use With (Equipment)Microscope slides
Slide Count20
Depth (Metric)6 mm
Diameter (Metric)30 mm
Unit SizeEA