Nalgene™ Polypropylene, Barbed Bulkhead Fittings, 6.5 mm
Nalgene™ Polypropylene, Barbed Bulkhead Fittings, 6.5 mm
Actual product may vary
Nalgene™ Polypropylene, Barbed Bulkhead Fittings, 6.5 mm
For Use With: Nalgene Bottle and Carboy Closures
Have Questions?
Catalog NumberQuantity
FKS12897650Case of 24
Catalog number FKS12897650
Price (MYR)
2,635.00
PK
Add to cart
Quantity:
Case of 24
Price (MYR)
2,635.00
PK
Add to cart
Customize tubing sets as culture vessels for aseptic transfer of liquids with autoclavable Thermo Scientific™ Nalgene™ Barbed Polypropylene Bulkhead Fittings, suitable for use with most Nalgene bottle and carboy closures. Pre-drilled 38mm-430 closures allow easy, safe in-house assembly.
Features:
  • 53B closures accept (two) 0.25″ fittings
  • Fittings, barbed at both ends, accept tubing for inside and outside container
Includes:
Instructions, drilling template and two each of polypropylene barbed fitting, acetyl nuts, silicone gaskets and and TPE port caps.
Specifications
Diameter (English) Inner Tubing0.25 in.
Diameter (Metric) Inner Tubing6.5 mm
For Use With (Equipment)Nalgene bottles and carboys
MaterialPolypropylene
No. per Case24/Cs.
No. per Pack2 Pk.
Product TypeFitting
QuantityCase of 24
Unit SizePK