Borosilicate Glass Automatic Burette
Borosilicate Glass Automatic Burette
Actual product may vary
Fisherbrand
Borosilicate Glass Automatic Burette
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Type
FKS1289209410 mLAutomatic Buret w/ Schellbach Stripe
FKS1288209450 mLAutomatic Buret
Catalog number FKS12892094
Price (MYR)
2,678.00
EA
Add to cart
Capacity (Metric):
10 mL
Type:
Automatic Buret w/ Schellbach Stripe
Price (MYR)
2,678.00
EA
Add to cart
  • With automatic zero facility
  • Simple operation - To fill burette apply a slight positive pressure to lower sidearm - To set zero, release pressure to allow excess liquid to siphon back into the bottle
  • Burettes manufactured from chemically resistant Pyrex™ borosilicate glass
  • White enamel graduations
  • 1.5mm bore PTFE key stopcock
  • The following accessories are sold separately: Glass Reservoir (3297/12) and Rubber Blowing Ball for Class AS Auto Burets (3297/14AS)
Specifications
Capacity (Metric)10 mL
MaterialChemically Resistant Pyrex Borosilicate Glass
GraduationsWhite Enamel at 0.02 mL
Includes1.5 mm bore PTFE key stopcock
TypeAutomatic Buret w/ Schellbach Stripe
Certifications/ComplianceISO 385 and DIN 12700
Unit SizeEA