Check Order Status
UV Transilluminator 2020M
Actual product may vary
UV Transilluminator 2020M
UV Transilluminator 2020M is for illumination of any sample excited by UV light.
Have Questions?
Catalog Number
Wavelength
FKS12874028
302 nm
Catalog number
FKS12874028
Price (MYR)
16,351.00
EA
Add to cart
Wavelength:
302 nm
Price (MYR)
16,351.00
EA
Add to cart
Sealed to allow wet gels on surface
For use in Syngene Systems
Specifications
Product Type
Single UV Transilluminator
Voltage
100/115/220/230 V
Intensity
Fixed
Wavelength
302 nm
Lighting Type
UV