Search Thermo Fisher Scientific
| Catalog Number | Capacity (Metric) | Bottom Shape |
|---|---|---|
| FKS12861541 | 2000 mL | Conical |
| FISK953901-0502 | 500 mL | Conical |
| FKI953901-0502 | 500 mL | Conical |
| FISK953901-2002 | 2000 mL | Conical |
| FKIK953901-2002 | 2000 mL | Conical |