Soda Lime Glass Bottle with 28mm Fitted Closure, Clear
Soda Lime Glass Bottle with 28mm Fitted Closure, Clear
Actual product may vary
Fisher Catalogue
Soda Lime Glass Bottle with 28mm Fitted Closure, Clear
Use the Fisherbrand™ Soda Lime Glass Bottle with 28mm Fitted Closure, Clear in the laboratory to store and manipulate laboratory media for experimentation.
Have Questions?
Catalog NumberCapacity (Metric)
FKS128608841000 mL
Catalog number FKS12860884
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
  • Tamper evident vistop closure with polycone liner
  • ISO certified
  • Clear soda lime glass
Specifications
Capacity (Metric)1000 mL
ColorClear
Diameter (Metric) Inner97.5 mm
Fill Volume Range1008mL
MaterialSoda Lime Glass
Neck ID (Metric)28mm
No. per Pack10 Pack
Quantity10/Pk
Screw MaterialPolypropylene
CertifiedISO
Closure ColorWhite
Liner MaterialEPE
Closure IncludedYes
ShapeRound
Height (Metric)215.6 mm
Product TypeLaboratory Bottle
Closure LinerPolypropylene/SOR
Closure MaterialPolypropylene
Diameter (Metric)97.5 mm
Unit SizePK