Use these cabinets for educational uses, viewing minerals and other applications. UVP™ C-15G Mini Chromato-Vue™ UV Viewing Cabinets provide a darkroom environment with UV fluorescence for viewing materials.
Have Questions?
Catalog Number
Voltage
FKS12823838
230V
Catalog number FKS12823838
Price (MYR)
10,556.00
EA
Add to cart
Voltage:
230V
Price (MYR)
10,556.00
EA
Add to cart
Designed with a metal base and plastic top
Includes a UVGL-58 UV lamp
The bottom panel of the cabinet is removable
The lamp fits snugly on top of the cabinet
A contrast control filter absorbs UV energy and protects the eyes
Soft rubber viewport provides a comfortable viewing area with space for eyeglasses
Lightweight access curtain blocks out external light from the cabinet interior
Lamps for other mini cabinets are ordered separately
Specifications
Voltage230V
Depth (Metric)229mm
Height (Metric)305mm
Width (Metric)267mm
ModelC-15G
Packaging Quantity1Pack
For Use With (Application)Mineralogy, Lab Fluorescence, Sterilization, Field Investigation