Test Tube Brushes with Bristle Tuft End
Test Tube Brushes with Bristle Tuft End
Actual product may vary
Fisherbrand
Test Tube Brushes with Bristle Tuft End
With bristle tuft end. Pinnacle Brush™. Test Tube Brushes with Bristle Tuft End is for the cleaning of test tubes.
Have Questions?
Catalog NumberLength (Metric)
FKS12801964265 mm
Catalog number FKS12801964
Price (MYR)
-
Length (Metric):
265 mm
  • Galvanised steel wire handle
  • With Bristle Tuft End
Specifications
Product TypeTest Tube Brush
Handle MaterialSteel Wire
For Use With (Equipment)Test tubes
Length (Metric)265 mm
Quantity1 Ea.
Size50 x 45 mm
Unit SizeEA