Clear Soda Lime Glass Bottle with Fitted Closure
Clear Soda Lime Glass Bottle with Fitted Closure
Actual product may vary
Fisherbrand
Clear Soda Lime Glass Bottle with Fitted Closure
Use the Fisherbrand™ Soda Lime Glass Bottle with 28mm Fitted Closure, Clear in the laboratory to store and manipulate laboratory media for experimentation.
Have Questions?
Catalog NumberCapacity (Metric)
FKS12800894100 mL
Catalog number FKS12800894
Price (MYR)
191.00
PK
Add to cart
Capacity (Metric):
100 mL
Price (MYR)
191.00
PK
Add to cart
  • Tamper evident vistop closure with polycone liner
  • ISO certified
  • Clear soda lime glass
Specifications
Capacity (Metric)100 mL
ColorClear
Diameter (Metric) Inner46.1 mm
MaterialGlass (Type 3)
Neck ID (Metric)28mm
No. per Pack20Pack
Quantity20 Pack
Closure ColorWhite
Liner MaterialLDPE
Closure IncludedYes
ShapeRound
Height (Metric)104.8 mm
Product TypeLaboratory Bottle
Closure MaterialHDPE
Diameter (Metric)46.1 mm
Closure TypeTamper-evident Vistop
Unit SizePK