Browne™ Plain Masking Tape
Browne™ Plain Masking Tape
Actual product may vary
Fisherbrand
Browne™ Plain Masking Tape
Use Browne™ Plain Masking Tape when autoclaving to hold objects into place and keep them from moving or sliding since the tape's efficacy is not affected by steam..
Have Questions?
Catalog NumberDimensions (L x W)Sterilization Indicator
FKS1278912550 m x 19 mmNo
Catalog number FKS12789125
Price (MYR)
-
Dimensions (L x W):
50 m x 19 mm
Sterilization Indicator:
No
  • Plain paper
  • Cream
Specifications
ColorCream
Dimensions (L x W)50 m x 19 mm
For Use With (Application)For process indicator autoclave
Product TypePlain Masking Tape
FeaturesNon-steam sensitive, Plain tape is similar to steam indicator tape but without the stripes
MaterialPaper
Quantity48 Carton
Sterilization IndicatorNo
Sterilization MethodSteam
Unit SizeEA