Borosilicate Glass Crystallizing Dish
Borosilicate Glass Crystallizing Dish
Actual product may vary
Fisherbrand
Borosilicate Glass Crystallizing Dish
Evaporate and crystallize substances in the laboratory using the Borosilicate Glass Crystallizing Dish without spout. his piece of laboratory glassware plays an important role in changing the properties of experimental substances. .
Have Questions?
Catalog NumberCapacity (Metric)
FKS12777316750 mL
Catalog number FKS12777316
Price (MYR)
114.00
EA
Add to cart
Capacity (Metric):
750 mL
Price (MYR)
114.00
EA
Add to cart
  • Borosilicate Glass
  • Flat bottom
  • Without spout
  • Capacity: 750mL
Specifications
Bottom ShapeFlat
Capacity (Metric)750 mL
MaterialBorosilicate Glass
ShapeRound
TypeCrystallizing Dish
Height (Metric)75 mm
Diameter (Metric)130 mm
Unit SizeEA