Cardboard Cryobox with Grid Insert And Lid
Cardboard Cryobox with Grid Insert And Lid
Actual product may vary
Fisherbrand
Cardboard Cryobox with Grid Insert And Lid
Highest flexibility: Cryoboxes can be combined with different grid inserts for tubes from 6 to 28.5mm in diameter
Have Questions?
Catalog number FKS12767125
Price (MYR)
444.00
PK
Add to cart
Price (MYR)
444.00
PK
Add to cart
  • Available with a water-repellent finish or with a waterproof plastic coating
  • Highest flexibility: Cryoboxes can be combined with different grid inserts for tubes from 6 to 28.5mm in diameter
  • The Cryoboxes can be used for larger vessels if the grid is removed
  • Ultra low temperature-resistant cardboard
Specifications
MaterialCardboard
ShapeSquare
TypeCryo Box
Tube Array10 x 10
Length (Metric)136 mm
Width (Metric)136 mm
Height (Metric)75 mm
Weight (Metric)1 kg
Unit SizePK