Check Order Status
Aluminum Basin with Lid
Actual product may vary
Fisherbrand
Aluminum Basin with Lid
Made of durable aluminum, these Nickel Electro™ Aluminum Basins with Lid are ideal in laboratory environments.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FKS12747356
45 mL
FKS12727346
10 mL
2 Options
Catalog number
FKS12747356
Price (MYR)
160.00
EA
Add to cart
Capacity (Metric):
45 mL
Price (MYR)
160.00
EA
Add to cart
For coal moisture determination
Flat, bottom aluminum basin with lid
Specifications
Bottom Shape
Flat
Capacity (Metric)
45 mL
Material
Aluminum
Type
Evaporating Basin
For Use With (Application)
For Coal Moisture Determination
Height (Metric)
14 mm
Diameter (Metric)
65 mm