Stainless Steel Rectangular Electric Boiling Baths
Stainless Steel Rectangular Electric Boiling Baths
Actual product may vary
Fisherbrand
Stainless Steel Rectangular Electric Boiling Baths
Get optimum boiling results with this rectangular, stainless steel electrically heated boiling bath. Featuring an energy regulator that controls the amount of boiling, this Clifton™ Stainless Steel Rectangular Electric Boiling Bath is ideal for safety.
Have Questions?
Catalog NumberModelDepth (Metric)Capacity (Metric)Temperature Range
FKS127188781104 E321 mm8.8 LConstant boiling at 100°C
Catalog number FKS12718878
Price (MYR)
9,744.00
EA
Add to cart
Model:
1104 E
Depth (Metric):
321 mm
Capacity (Metric):
8.8 L
Temperature Range:
Constant boiling at 100°C
Price (MYR)
9,744.00
EA
Add to cart
  • Rectangular, stainless steel electrically heated
  • Multi-place baths with fixed temperature for boiling water
  • Fitted with concentric ringed lids to vary the hole size
  • Features an integral constant level device to maintain constant water level during boiling
  • Fitted with a sheathed immersion element and a one shot thermal cut out for safety
Specifications
Controller TypeEnergy regulator controls the amount of boiling from a gentle simmer to vigorous boil
DescriptionBoiling Bath
Capacity (Metric)8.8 L
Temperature RangeConstant boiling at 100°C
For Use With (Application)Distilling/Concentrating Liquids, Melting Agar and Other Solids, Metallurgical Analysis, Protein Ana
MaterialStainless Steel
Model1104 E
No. of Positions12 x 80 mm Holes
ShapeRectangular
Warranty2 Years
Dimensions (D x W x H)321 x 425 x 90 mm
Width (Metric)425 mm
Height (Metric)90 mm
Depth (Metric)321 mm
Weight (Metric)5 kg