Nickel 201 Laboratory Dishes
Nickel 201 Laboratory Dishes
Actual product may vary
Nickel 201 Laboratory Dishes
Nickel Electro™ Nickel 201 Laboratory Dishes are made to resist corrosive enviroments, which make it useful for maintaining product purity.
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric)Height (Metric)
FKS12717366100 mL90 mm16 mm
Catalog number FKS12717366
Price (MYR)
509.00
EA
Add to cart
Capacity (Metric):
100 mL
Diameter (Metric):
90 mm
Height (Metric):
16 mm
Price (MYR)
509.00
EA
Add to cart
  • 99.6% nickel purity
  • Crevice free
  • High thermal and electrical conductivities
  • Low gas content
  • Low vapor pressure
  • Ideal for lab crucibles that are capable of withstanding oxidizing furnace atmospheres up to 1100°C
Specifications
Bottom ShapeFlat
Capacity (Metric)100 mL
MaterialNickel
ShapeRound
TypeEvaporating Dish
Interior FinishBright
Height (Metric)16 mm
Diameter (Metric)90 mm
Unit SizeEA