Check Order Status
Cellulose Dialysis Tubing Roll
Actual product may vary
Cellulose Dialysis Tubing Roll
Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies.
Have Questions?
Catalog Number
MWCO
Capacity (Metric)
FKS12707476
14 kDa
6.45 mL/cm
Catalog number
FKS12707476
Price (MYR)
1,848.00
EA
Add to cart
MWCO:
14 kDa
Capacity (Metric):
6.45 mL/cm
Price (MYR)
1,848.00
EA
Add to cart
Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies.
Seamless, thin, transparent dialysis tubing manufactured from the highest purity cellulose
Just wet in distilled water and use
Available in 30.5m rolls
Specially purified for life science research
Supplied in an airtight, moisture-proof container for maximum shelf life
Available in 14000Da, 8000Da, and 3500Da molecular weight cut offs
Specifications
Product Type
Dialysis Tubing Roll
MWCO
14 kDa
Material
Cellulose
Diameter (Metric)
28.7 mm
Length (Metric)
30.5 m
Capacity (Metric)
6.45 mL/cm
Color
Transparent
Quantity
1 Each
For Use With (Application)
For life sciences research