Browne™ Control™ Autoclave Indicator Valves
Browne™ Control™ Autoclave Indicator Valves
Actual product may vary
Fisherbrand
Browne™ Control™ Autoclave Indicator Valves
Test the efficacy of an autoclave using Browne™ Control™ Autoclave Indicator Valves. These tubes contain heat-sensitive dyes that change color after sufficient time at the autoclave's desired temperature.
Have Questions?
Catalog NumberColor
FKS12705335Black Spot
Catalog number FKS12705335
Price (MYR)
645.00
PK
Add to cart
Color:
Black Spot
Price (MYR)
645.00
PK
Add to cart
Test the efficacy of an autoclave using Browne™ Control™ Autoclave Indicator Valves. These tubes contain heat-sensitive dyes that change color after sufficient time at the autoclave's desired temperature.
  • The use of TST Control™ emulating Indicators is recommended for monitoring porous load steam sterilization processes
  • Various colors
  • Glass tubes
Specifications
ColorBlack Spot
For Use With (Application)For monitoring porous load steam sterilization processes
For Use With (Equipment)Autoclave
Product TypeBrowne Control Tube
Temperature (Metric)121°C
Quantity100 Pk.
Sterilization IndicatorYes
Sterilization Time15 min.
Sterilization MethodSteam, Dry Heat
Unit SizePK