Polystyrene Petri Dishes, Sterile
Polystyrene Petri Dishes, Sterile
Actual product may vary
Fisherbrand
Polystyrene Petri Dishes, Sterile
A range of high quality aseptic or sterile Petri dishes perfect for microbial or cell culture, or for sample collection, examination and storage.
Have Questions?
Catalog NumberHeight (Metric)Vent
FKS1267478516.2 mm1
Catalog number FKS12674785
Price (MYR)
1,024.00
PK
Add to cart
Height (Metric):
16.2 mm
Vent:
1
Price (MYR)
1,024.00
PK
Add to cart
Specifications
MaterialPolystyrene
SterilitySterile
TypePetri Dish
Quantity600 Case
Vent1
Height (Metric)16.2 mm
Diameter (Metric)90 mm
Unit SizePK