MB22 Premier Disposable Low-profile Microtome Blade
MB22 Premier Disposable Low-profile Microtome Blade
Actual product may vary
Fisherbrand
MB22 Premier Disposable Low-profile Microtome Blade
Choose the Epredia™ MB22 premier disposable low-profile microtome blade for applications where extremely thin, high-quality sections are required.
Have Questions?
Catalog NumberProfile
FKS12647896Low
Catalog number FKS12647896
Price (MYR)
1,170.00
PK
Add to cart
Profile:
Low
Price (MYR)
1,170.00
PK
Add to cart
  • Low-profile
  • Increased durability
Specifications
Product TypeLow Profile Blade
ProfileLow
For Use With (Application)Designed with a steeper angle that allows users to cut thinner sections
DisposableSingle-use
Cutting Angle22°
Thickness (English)0.01 in.
Thickness (Metric)0.25 mm
Unit SizePK