Works with a variety of aquaria with availability in a number of different sizes. Azpack™ Aquarium Air Pump powers aquarium air filters quietly and efficiently.
Have Questions?
Catalog Number
Flow Rate
Maximum Pressure
Model
Overall Length (Metric)
FKS12473170
3.5 L/min.
0.17 mbar
AP2
1 m
Catalog number FKS12473170
Price (MYR)
-
Flow Rate:
3.5 L/min.
Maximum Pressure:
0.17 mbar
Model:
AP2
Overall Length (Metric):
1 m
Reliable hard wearing pumps with anti-vibration feet