Wheaton™ Reusable PVC Coliwasas and Accessories
Wheaton™ Reusable PVC Coliwasas and Accessories
Actual product may vary
Wheaton™ Reusable PVC Coliwasas and Accessories
For a variety of applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeLength (Metric)Length (English)
FKS12437311Coliwasa Sampler1.82 m6 ft.
FKS11734129Extension and Connector1.21 m4 ft.
Catalog number FKS12437311
Price (MYR)
-
Product Type:
Coliwasa Sampler
Length (Metric):
1.82 m
Length (English):
6 ft.
Specifications
Product TypeColiwasa Sampler
MaterialPVC
Includes48 in. L cleaning brush
Capacity (English)37.19 oz.
Capacity (Metric)1100 mL
Diameter (English) Inner1.37 in.
Diameter (English) Outer1.61 in.
Diameter (Metric) Inner3.5 cm
Diameter (Metric) Outer4.1 cm
Length (English)6 ft.
Length (Metric)1.82 m
Unit SizeEA