Mortar and Pestle with Wooden Handle Pestle
Mortar and Pestle with Wooden Handle Pestle
Actual product may vary
Fisherbrand
Mortar and Pestle with Wooden Handle Pestle
Releases oils that can't be extracted using modern appliances. Azpack™ Mortar and Pestle with Wooden Handle will grind and crush the toughest materials without cracking the strong and dense porcelain.
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric) InnerHeight (Metric)Length (Metric)
FKS12393338199 mL95mm74mm (Mortar)170mm (Pestle)
Catalog number FKS12393338
Price (MYR)
-
Capacity (Metric):
199 mL
Diameter (Metric) Inner:
95mm
Height (Metric):
74mm (Mortar)
Length (Metric):
170mm (Pestle)
  • Mortar constructed of vitrified electical porcelain
  • Pestle includes a Beech wood handle
  • Unglazed grinding surface
  • Integral pouring lip reduces waste and spillage
Specifications
Capacity (Metric)199 mL
FinishUnglazed
Diameter (Metric) Inner95mm
Diameter (Metric) Outer113mm (118mm to lip)
Product TypeMortar and Pestle
MaterialMortar: Vitrified electrical porcelain, Pestle: Beech
Height (Metric)74mm (Mortar)
Length (Metric)170mm (Pestle)
Unit SizeEA