Check Order Status
Sterile Microcentrifuge Tubes with Screw Caps
Actual product may vary
Sterile Microcentrifuge Tubes with Screw Caps
Note:
Image is representative only. It shows colored caps, but all of the tubes in this offering come with Natural caps; there are no color cap options.
Have Questions?
Catalog Number
Capacity (Metric)
Max. RCF
dimShape
Bottom Shape
FKS12390433
2 mL
25,000 x g
Conical
Catalog number
FKS12390433
Price (MYR)
1,060.00
PK
Add to cart
Capacity (Metric):
2 mL
Max. RCF:
25,000 x g
Bottom Shape:
Conical
Price (MYR)
1,060.00
PK
Add to cart
Choose skirted or conical-bottom style
With attached caps
Natural-colored screw caps with O-rings
RNase-free and DNase-free
Specifications
Closure Material
Polypropylene
Autoclavable
Autoclavable
Capacity (Metric)
2 mL
Color
Natural
Material
Polypropylene
Includes
O-ring
Sterility
Sterile
Closure Type
Screw Cap
Bottom Shape
Conical
Max. RCF
25,000 x g