Check Order Status
Lids For Porcelain Crucibles
Actual product may vary
Lids For Porcelain Crucibles
Designed to fit Haldenwanger™ Porcelain Crucibles.
Have Questions?
Catalog Number
For Use With (Application)
FKS12386487
13mL crucibles
Catalog number
FKS12386487
Price (MYR)
965.00
PK
Add to cart
For Use With (Application):
13mL crucibles
Price (MYR)
965.00
PK
Add to cart
Available in a variety of sizes
For use with porcelain crucibles
Specifications
Color
White
Material
Porcelain
For Use With (Application)
13mL crucibles
Diameter (Metric) Outer
43mm
Product Type
Crucible Lids