Clear Glass Wide Neck Powder Bottles with Black Urea Screw Caps
Clear Glass Wide Neck Powder Bottles with Black Urea Screw Caps
Actual product may vary
Fisher Catalogue
Clear Glass Wide Neck Powder Bottles with Black Urea Screw Caps
Fill, dispense, and store powders and liquids without difficulty. Azpack™ Clear Glass Wide Neck Powder Bottles with Black Urea Screw Caps feature a wood pulp-faced PVDC-lined cap.
Have Questions?
Catalog NumberCapacity (Metric)Closure Size
FKS1237712830 mL33/R3
Catalog number FKS12377128
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
30 mL
Closure Size:
33/R3
  • Constructed of clear soda lime glass
  • Includes black urea screw cap closure
  • Closure lined with wood pulp faced steran (PVDC)
Specifications
Capacity (Metric)30 mL
ColorClear
MaterialSoda Lime Glass
Quantity40/PK
Closure ColorBlack
Closure IncludedYes
MouthWide, Threaded
Height (Metric)59.5 mm
Closure LinerWood Pulp-faced PVDC
Closure MaterialUrea
Closure Size33/R3
Diameter (Metric)39.7 mm
Closure TypeScrew Cap
Unit SizePK