Disposable Pipette Basins
Disposable Pipette Basins
Actual product may vary
Disposable Pipette Basins
Disposable Pipette Basins
Fisherbrand
Disposable Pipette Basins
Disposable basins offer convenience, fast cleanup.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)SterilityPackaging
FKS1236917550 mLNon-sterileBulk
FKS12399175100 mLSterileBulk
Catalog number FKS12369175
Price (MYR)
128.00
PK
Add to cart
Capacity (Metric):
50 mL
Sterility:
Non-sterile
Packaging:
Bulk
Price (MYR)
128.00
PK
Add to cart
Fisherbrand™ Disposable Pipette Basins are packaged in easy-to-open bags to reduce contamination risks. They offer convenience and fast cleanup.
  • Easy-pour corners for quick disposal of excess sample solution
25mL Well Basins
  • Dimensions: 5.7L × 2.4W × 0.8 in.H (14.5 × 6.3 × 2.1cm)
50mL Basins
  • Angled sides and V-shaped bottom permit easy withdrawal of sample solution
  • Basin is constructed of clear PVC; Sterile (unless opened)
  • Dimensions: 5.75L × 2.75W × 1 in.H (14.0 × 7 × 2.5cm)
100mL Basins
  • Versatile design allows ample clearance for all types of 4-, 8-, and 12-channel pipetters
  • Ideal for extended multichannel pipetting into microtiter plates, tubes, and deep-well blocks
  • Dimensions: 5.75L × 3.5W × 1.25 in.H (14.6 × 9 × 3.2cm)
Specifications
ColorClear
Capacity (Metric)50 mL
Dimensions (L x W x H)5.75 x 2.75 x 1 in. (14.6 x 7 x 2.54cm)
MaterialPVC
SterilityNon-sterile
TypePipetting Reservoir
Height (English)1 in.
Height (Metric)2.54 cm
Length (English)5.75 in.
Length (Metric)14.6 cm
PackagingBulk
Width (English)2.75 in.
Width (Metric)7 cm