Amber Glass Wide Neck Powder Bottles with Black Urea Screw Caps
Amber Glass Wide Neck Powder Bottles with Black Urea Screw Caps
Actual product may vary
Fisherbrand
Amber Glass Wide Neck Powder Bottles with Black Urea Screw Caps
Use these bottles for general storage applications. Azpack™ Amber Glass Wide Neck Powder Bottles with Black Urea Screw Caps provide protection for light sensitive liquids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FKS12367148250 mL48/R3
FKS1231714830 mL33/R3
FKS1232714860 mL38/R3
FKS12347148125 mL38/R3
Catalog number FKS12367148
Price (MYR)
461.00
PK
Add to cart
Capacity (Metric):
250 mL
Closure Size:
48/R3
Price (MYR)
461.00
PK
Add to cart
  • Constructed of amber-colored soda lime glass
  • Includes black urea screw cap closure
  • Closure lined with wood pulp faced steran (PVDC)
Specifications
Capacity (Metric)250 mL
ColorAmber
MaterialSoda Lime Glass
Quantity20/PK
Closure ColorBlack
Closure IncludedYes
MouthWide
Height (Metric)121 mm
Closure LinerWood Pulp-faced PVDC
Closure MaterialUrea
Closure Size48/R3
Diameter (Metric)64 mm
Closure TypeScrew Cap
Unit SizePK