Check Order Status
Lids for Porcelain Crucibles
Actual product may vary
Lids for Porcelain Crucibles
Use these lids with porcelain crucibles. Haldenwanger™ Lids for Porcelain Crucibles are constructed from optimized laboratory porcelain.
Have Questions?
Catalog Number
Diameter (Metric) Outer
FKS12356507
35mm
Catalog number
FKS12356507
Price (MYR)
848.00
PK
Add to cart
Diameter (Metric) Outer:
35mm
Price (MYR)
848.00
PK
Add to cart
Available in a variety of sizes
For use with Haldenwanger porcelain crucibles
Complies with DIN VDE 0335, type C 100
Specifications
Material
Porcelain
Diameter (Metric) Outer
35mm
Product Type
Crucible Lid