Glass Square Coverslips
Glass Square Coverslips
Actual product may vary
Fisherbrand
Glass Square Coverslips
Glass Square Coverslips are available in a variety of sizes. Packed in plastic boxes to prevent breakage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberThickness (Metric)Length (Metric)Width (Metric)
FKS12333128#1 (0.13 to 0.16 mm)22 mm22 mm
FKS12393138#1.5 (0.16 to 0.19 mm)24 mm24 mm
Catalog number FKS12333128
Price (MYR)
33.00
EA
Add to cart
Thickness (Metric):
#1 (0.13 to 0.16 mm)
Length (Metric):
22 mm
Width (Metric):
22 mm
Price (MYR)
33.00
EA
Add to cart
  • Manufactured from clear soda glass
  • Consistent in cut, thickness and dimensional accuracy
  • Free from blisters and flaws
Specifications
Product TypeCoverslip
ShapeSquare
MaterialSoda Lime Glass
Length (Metric)22 mm
Width (Metric)22 mm
Thickness (Metric)#1 (0.13 to 0.16 mm)
Quantity200/Pk.
Unit SizeEA