Glass Circle Coverslips
Glass Circle Coverslips
Actual product may vary
Fisherbrand
Glass Circle Coverslips
Glass Circle Coverslips are available in two thicknesses and a variety of diameters. Packed in plastic boxes to prevent breakage.
Have Questions?
Catalog NumberThickness (Metric)Diameter (Metric)
FKS12313138#1 (0.13 to 0.16 mm)16 mm
Catalog number FKS12313138
Price (MYR)
78.00
PK
Add to cart
Thickness (Metric):
#1 (0.13 to 0.16 mm)
Diameter (Metric):
16 mm
Price (MYR)
78.00
PK
Add to cart
These Glass Circle Coverslips are available in two thicknesses and a variety of diameters. Packed in plastic boxes to prevent breakage.
  • Manufactured from clear soda glass
  • Consistent in cut, thickness and dimensional accuracy
  • Free from blisters and flaws
Specifications
Product TypeCoverslip
ShapeCircle
MaterialSoda Lime Glass
Thickness (Metric)#1 (0.13 to 0.16 mm)
Diameter (Metric)16 mm
Quantity200/Pk.
Unit SizePK