Check Order Status
Cylinder Brush
Actual product may vary
Fisherbrand
Cylinder Brush
Manufactured with galvanized steel handle. Pinnacle Brush™ Cylinder Brush features nylon bristles.
Have Questions?
Catalog Number
Length (Metric)
FKS12303745
685 mm
Catalog number
FKS12303745
Price (MYR)
-
Length (Metric):
685 mm
Fan-shaped end
Galvanised steel wire handle
Specifications
Product Type
Cylinder Brush
Bristle Material
Nylon
Handle Material
Steel Wire
Diameter (Metric) Bristle
64 mm
Length (Metric)
685 mm
Length (Metric) Brush
150 mm
Quantity
10 Pk.
Size
64 x 150 mm