Check Order Status
Plastic Straight Hose Connector
Actual product may vary
Plastic Straight Hose Connector
For connecting screw caps with bore. Lenz Laborglasinstrumente™ Plastic Straight Hose Connectors are made from polypropylene.
Have Questions?
Catalog Number
Thread Size
FKS12303379
GL18
Catalog number
FKS12303379
Price (MYR)
763.00
PK
Add to cart
Thread Size:
GL18
Price (MYR)
763.00
PK
Add to cart
Autoclavable
Available in two different thread sizes
Silicone sealing
Specifications
Autoclavable
Autoclavable
Material
Polypropylene
For Use With
For screw caps with bore
Packaging Quantity
10 Pack
Thread Size
GL18
Shape
Straight
Product Type
Straight Hose Connection