0.2mL PCR Tubes
0.2mL PCR Tubes
Actual product may vary
0.2mL PCR Tubes
With integral snap shut cap
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorClosure Type
FKS12134102NaturalFlat Cap
FIS14-230-205NaturalDomed Cap
FIS14-230-225NaturalFlat Cap
Catalog number FKS12134102
Price (MYR)
429.00
PK
Add to cart
Color:
Natural
Closure Type:
Flat Cap
Price (MYR)
429.00
PK
Add to cart
With integral snap shut cap
  • Suitable for 0.2mL thermal cycler blocks
  • 0.25mL maximum volume when closed
  • Thin-wall design
  • Cleanroom produced
  • Virgin polypropylene
Specifications
Certifications/ComplianceRNase, Dnase and human DNA free
ColorNatural
Volume (Metric)0.2 mL
Quantity1000 Case
For Use With (Equipment)0.2mL thermal cycler blocks for PCR and sample storage applications
MaterialPolypropylene
Closure TypeFlat Cap
FormatTube
Product TypeThin Wall PCR Tube
Unit SizePK