Check Order Status
0.2mL PCR Tube Strips
Actual product may vary
0.2mL PCR Tube Strips
Ideal for use in 0.2mL, 96-well V-bottom thermal cyclers.
Have Questions?
Change view
Catalog Number
Color
Closure Type
FKS12104092
Natural
Domed Cap
FIS14-230-210
Natural
Domed Cap
FIS14-230-215
Natural
Flat Cap
3 Options
Catalog number
FKS12104092
Price (MYR)
1,645.00
PK
Add to cart
Color:
Natural
Closure Type:
Domed Cap
Price (MYR)
1,645.00
PK
Add to cart
Ideal for use in 0.2mL, 96-well V-bottom thermal cyclers
0.25mL maximum tube volume when capped
Strips of eight tubes
Can be cut into individual tubes
Thin-wall design
Cleanroom produced
Virgin polypropylene
Available with flat cap strips, domed cap strips, or without cap strips
Specifications
Certifications/Compliance
Human DNA, RNase, DNase, endotoxins free
Color
Natural
Volume (Metric)
0.2 mL
Quantity
250 Case
For Use With (Equipment)
0.2mL 96-well V-bottom thermal cyclers
Material
Polypropylene
Closure Type
Domed Cap
No. of Wells
96
Format
Strip
Product Type
Thin Wall 8-Strip PCR Tube