Check Order Status
Pyrex™ Filter Holder Support Base
Actual product may vary
Fisherbrand
Pyrex™ Filter Holder Support Base
Pyrex™ Filter Holder Support Base is a key component used in the assembly of the Pyrex™ 47 mm Membrane Filtration System.
Have Questions?
Catalog number
FKS11997098
Price (MYR)
3,834.00
EA
Add to cart
Price (MYR)
3,834.00
EA
Add to cart
Pyrex™ Filter Holder Support Base is a key component used in the assembly of the Pyrex™ 47 mm Membrane Filtration System.
Support base with sintered disc and vacuum connection with 40/38 socket
Designed to be connected below the filtration system funnel with a clamp
Once connected to the funnel, place base on top of the opening to the flask
For use with the all glass filtration system
Borosilicate glass porosity grade 3 sintered disc to support membrane
Fully autoclavable
Ideal for microbiology and preparation of HPLC solvents
Clamp, funnel, and flask sold separately
Specifications
Material
Borosilicate Glass
For Use With (Application)
For microbiology applications
Includes
Sintered disc and vacuum connection
Porosity
Grade 3
Type
Support Base
Diameter (Metric) Disc
47mm