Narrow-Mouth Field Sample Bottles
Narrow-Mouth Field Sample Bottles
Actual product may vary
Narrow-Mouth Field Sample Bottles
Narrow-Mouth Field Sample Bottles
Narrow-Mouth Field Sample Bottles
Narrow-Mouth Field Sample Bottles
Bulk-packaged for convenient liquid sampling, transport, and storage
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS119779241000 mL
FIS02-895A125 mL
FIS02-895B250 mL
FIS02-895D1000 mL
Catalog number FKS11977924
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Bulk-packaged for convenient liquid sampling, transport, and storage
  • HDPE for excellent chemical resistance
  • Rigid and translucent
  • Ideal for field sampling
Specifications
Capacity32 oz., 1000 mL
Capacity (Metric)1000 mL
Capacity (English)32 oz.
ColorTranslucent
MaterialHDPE
Neck Finish38-430
Certifications/ComplianceISO 13485:2003; BSI:FM 653694; QMI-SAI:File No.1606319 and 1606321
Closure IncludedYes
MouthNarrow
Product TypeNarrow Mouth Bottle
Closure MaterialPolypropylene
Closure Size38-430 mm
Closure TypeScrew Cap
Unit SizeEA