▸
Facility Safety and Maintenance
▸
Cleaning Supplies and Equipment
▸
Odor Control Products
▸
OdoClave™ Deodorant Autoclave Pads
Actual product may vary
Fisherbrand
OdoClave™ Deodorant Autoclave Pads
Neutralize autoclave odors with a pleasant fragrance
Have Questions?
Change view
Catalog Number
Scent
Color
FKS11976072
Clove
Brown
FKS11956072
Cherry
Red
2 Options
Catalog number
FKS11976072
Price (MYR)
2,071.00
Pack
Add to cart
Scent:
Clove
Color:
Brown
Price (MYR)
2,071.00
Pack
Add to cart
Neutralize autoclave odors with a pleasant fragrance
Heat-release deodorant works automatically at autoclave temperatures
Open pouch and place in bag prior to autoclaving
Three-year shelf life or longer if stored in a sealed container
32mm (1.75 in.) square
Triple strength pads release three times the deodorant of standard pads while costing less than two standard pads
Specifications
Product Type
Autoclave Capsule
Color-coded
No
Form
Liquid
Scent
Clove
Sterility
Non-sterile
Autoclavable
Autoclavable
Color
Brown
Format
Pad
For Use With (Application)
Autoclave