Glass Pear-Shaped Separating Funnel with PTFE Stopcock
Glass Pear-Shaped Separating Funnel with PTFE Stopcock
Actual product may vary
Fisherbrand
Glass Pear-Shaped Separating Funnel with PTFE Stopcock
Constructed in chemical resistant borosilicate glass. Fisherbrand™ Glass Pear-Shaped Separating Funnel with PTFE Stopcock separates mixtures into immiscible solvents. Available with interchangeable glass or PTFE stopcock, but supplied with a glass and plastic stopper.
Have Questions?
Catalog NumberCapacity (Metric)Stopper Number
FKS11972138500 mL24/29
Catalog number FKS11972138
Price (MYR)
723.00
EA
Add to cart
Capacity (Metric):
500 mL
Stopper Number:
24/29
Price (MYR)
723.00
EA
Add to cart
Constructed in chemical resistant borosilicate glass. Fisherbrand™ Glass Pear-Shaped Separating Funnel with PTFE Stopcock separates mixtures into immiscible solvents. Available with interchangeable glass or PTFE stopcock, but supplied with a glass and plastic stopper.
  • Constructed in chemical resistant borosilicate glass.
  • ISO 4800 Certified.
    • Easy-open stopper helps to reduce pressure buildup during processing.
    Specifications
    Capacity (Metric)500 mL
    MaterialBorosilicate Glass
    Certifications/ComplianceISO 4800
    Quantity1PK
    StopcockPTFE
    Stopper MaterialGlass, Plastic
    Stopper Number24/29
    Product TypeSeparatory Funnel
    Unit SizeEA