Pyrex™ Borosilicate Glass General Purpose Dish
Pyrex™ Borosilicate Glass General Purpose Dish
Actual product may vary
Fisherbrand
Pyrex™ Borosilicate Glass General Purpose Dish
Autoclavable and chemically resistant
Have Questions?
Catalog NumberCapacity (Metric)
FKS119425482000 mL
Catalog number FKS11942548
Price (MYR)
-
Capacity (Metric):
2000 mL
Autoclavable and chemically resistant
  • Manufactured from chemically resistant Pyrex™ borosilicate glass
  • Dishes can be autoclaved or used in microwave ovens
Specifications
Capacity (Metric)2000 mL
Dimensions (L x W x H)275 x 200 x 51 mm
Height (Metric)57 mm
Length (Metric)262 mm
MaterialGlass
TypeGeneral Purpose Dish
Width (Metric)199 mm
For Use WithMicrowave ovens
For Use With (Application)Ideal for drying powders and crystals, staining and development, collection of drips and spills from solution bottles, gel electrophoresis and paper chromatography dissection work
Length (English)10.31 in.
Height (English)2.24 in.
Resistant ToChemicals
ShapeRectangular
Width (English)7.83 in.