Check Order Status
Cup Sampler
Actual product may vary
Cup Sampler
Manufactured from 316 stainless steel. The Cup Sampler enables discrete samples to be easily taken from known heights within the product.
Have Questions?
Catalog Number
Capacity (Metric)
FKS11940018
100 mL
Catalog number
FKS11940018
Price (MYR)
5,392.00
EA
Add to cart
Capacity (Metric):
100 mL
Price (MYR)
5,392.00
EA
Add to cart
Key Features:
Single point liquid sampler
Simple and easy operation
All product contact parts are made from 316L Stainless Steel
Variety of lengths and cup sizes available
Operation:
Insert the sampler to the required depth.
Squeeze handle which pushes the cup away from the lid, allowing the liquid to enter the cup.
Once the sample has been taken release the handle and remove from the product.
To empty the sampler either tip the sampler whilst squeezing the handle or unscrew the collecting cup.
Specifications
Product Type
Cup Sampler
Material
Stainless Steel (316)
Capacity (Metric)
100 mL
Disposable
Reusable
Diameter (Metric) Outer
55 mm
Length (Metric)
1500 mm