Carbon Steel Razor Blades
Actual product may vary
Fisherbrand
Carbon Steel Razor Blades
Packaged in drawer box merchandiser. Azpack™ Carbon Steel Razor Blades are individually wrapped in paper cardstock for safety.
Have Questions?
Catalog NumberSize
FKS1190432538.1 x 19 mm
Catalog number FKS11904325
Price (MYR)
477.00
Each
Size:
38.1 x 19 mm
Price (MYR)
477.00
Each
Packaged in drawer box merchandiser. Azpack™ Carbon Steel Razor Blades are individually wrapped in paper cardstock for safety.
  • Carbon steel
  • Regular duty
  • Single-edged blade
  • Contains aluminum spine
  • Individually wrapped
Specifications
Blade MaterialCarbon Steel
Product TypeRazor Blade
Size38.1 x 19 mm
Thickness (Metric)0.22 mm
Quantity10/Pk.
Unit SizeEach