Constructed of sturdy polypropylene. Test Tube Racks include alphanumeric positions for easy sample identification. Racks are stackable, non-conductive, and will not float in water baths.
Have Questions?
Catalog Number
Color
Holds
FKS11889550
White
Tubes (16 mm)
Catalog number FKS11889550
Price (MYR)
796.00
PK
Add to cart
Color:
White
Holds:
Tubes (16 mm)
Price (MYR)
796.00
PK
Add to cart
Sturdy polypropylene racks include alphanumeric positions for easy sample identification.
Racks are stackable, non-conductive and will not float in water baths.
Suitable for use in temperatures from -20° C to 90° C.
Supplied in three pieces for easy and permanent assembly.