Check Order Status
Cryopreservation Vials, Canes and Canisters
Actual product may vary
Cryopreservation Vials, Canes and Canisters
Maximize storage capacities in the laboratory by using these Cryopreservation Vials, Canes and Canisters.
Have Questions?
Catalog Number
Type
For Use With (Equipment)
FKS11884760
Cryo Cane
2 Ampules
Catalog number
FKS11884760
Price (MYR)
83.00
EA
Add to cart
Type:
Cryo Cane
For Use With (Equipment):
2 Ampules
Price (MYR)
83.00
EA
Add to cart
Maximize storage capacities using Thermo Scientific™ Cryopreservation Vials, Canes and Canisters.
Warranty varies by country, please contact us for details
Specifications
Description
Aluminum
Type
Cryo Cane
Capacity (Metric)
5 mL
Color
Silver
For Use With (Equipment)
2 Ampules
Holds
Ampules (2)